Learn More
Medchemexpress LLC Tau-D/0N4R Human 1 5ug
SDP
Catalog No. 5000196774
Encompass_Preferred
Recombinant fragment of human microtubule-associated protein tau (residues Q249-Q381), expressed in Escherichia coli with a C-terminal 6xHis tag. The 133-amino-acid protein is supplied at approximately 16-18 kDa and ≥95% purity in a 5 μg format for biochemical and neurodegeneration research applications.
  • Recombinant human tau fragment covering Q249-Q381.
  • C-terminal 6xHis tag for purification and detection.
  • Expressed in Escherichia coli to enable reproducible production.
  • High purity suitable for biochemical and binding assays.
  • Approximate molecular weight 16-18 kDa for gel confirmation.
  • Supplied in a 5 μg format suitable for assay development.
Catalog No. 50-001-96774 Supplier Medchemexpress LLC Supplier No. HYP711025UG
Recombinant fragment of human microtubule-associated protein tau (residues Q249-Q381), expressed in Escherichia coli with a C-terminal 6xHis tag. The 133-amino-acid protein is supplied at approximately 16-18 kDa and ≥95% purity in a 5 μg format for biochemical and neurodegeneration research applications.
  • Recombinant human tau fragment covering Q249-Q381.
  • C-terminal 6xHis tag for purification and detection.
  • Expressed in Escherichia coli to enable reproducible production.
  • High purity suitable for biochemical and binding assays.
  • Approximate molecular weight 16-18 kDa for gel confirmation.
  • Supplied in a 5 μg format suitable for assay development.