Learn More
Enzo Life Sciences Tau (human) monoclonal antibody (Tau 12) (0.1ml)
SDP
Supplier:  Enzo Life Sciences ENZABS2160100
Encompass_Preferred
Clone: Tau 12. Isotype: IgG1. Species reactivity: Human. Specificity: Recognizes an epitope between aa 6-18 of human Tau. Applications: ELISA, IHC, WB. Purity: Purified. Formulation: Liquid. In PBS. Does not contain any preservatives or carrier proteins.
Catalog No. 50-201-2765