Learn More
Medchemexpress LLC Tau peptide (273-284) | 1663490-91-2 | 98.8% | 1368.66 | 10 MG
SDP
Catalog No. 5000397129
Encompass_Preferred
Tau Peptide (273-284) is a Tau fragment designed for research in tauopathies, particularly Alzheimer's disease. This peptide is for research use only and not intended for patient treatment. In vitro studies show that Tau Peptide (273-284) (50 μM) forms β-structures faster in the ΔK280 mutant compared to its wild-type counterpart.
  • Used in research of tauopathies
  • Forms β-structures faster in ΔK280 mutant
  • White to off-white solid appearance
  • Available in various pack sizes
Catalog No. 50-003-97129 Supplier Medchemexpress LLC Supplier No. HYP494310MG
Tau Peptide (273-284) is a Tau fragment designed for research in tauopathies, particularly Alzheimer's disease. This peptide is for research use only and not intended for patient treatment. In vitro studies show that Tau Peptide (273-284) (50 μM) forms β-structures faster in the ΔK280 mutant compared to its wild-type counterpart.
  • Used in research of tauopathies
  • Forms β-structures faster in ΔK280 mutant
  • White to off-white solid appearance
  • Available in various pack sizes