Learn More
Bachem Tau Peptide (298-312) Trifluoroacetate
SDP
Supplier:  Bachem 4099422.1000
Encompass_Preferred
Tau Peptide (298-312) Trifluoroacetate, H-Lys-His-Val-Pro-Gly-Gly-Gly-Ser-Val-Gln-Ile-Val-Tyr-Lys-Pro-OH, 330456-47-8, C72H116N20O19, Mr 1565.84, 1mg.
Catalog No. 50-194-6637