Learn More
CPC Scientific Ac-Gln-Thr-Ala-Pro-Val-Pro-Met-Pro-Asp-Leu-Lys-Asn-Val-Lys-Ser-Lys-Ile-Gly-Ser-Thr-Glu-Asn-Leu-Lys-His-Gln-Pro-Gly-Gly-Gly-Lys-OH (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific TAUP003B
Encompass
TAU proteins belong to the microtubule-associated protein (MAP) family and are involved in the pathogenesis of Alzheimer?s disease. In the human brain, there are six TAU isoforms ranging from 352 to 441 amino acids in length. These isoforms vary at the carboxyl terminal according to the presence of either three repeat or four repeat domains (R1-R4), in addition to the presence or absence of one or two insert domains at the amino-terminus (see figure below). Tau Peptide (244-274) is a 31-amino acid long peptide derived from the Repeat 1 domainONE-LETTER SEQUENCE: Ac-QTAPVPMPDLKNVKSKIGSTENLKHQPGGGKMOLECULAR FORMULA: C143H240N42O45S1MOLECULAR WEIGHT:3299.80STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [2022956-56-3], SYNONYMS: Acetyl-Tau Peptide (244-274) (Repeat 1 Domain)RESEARCH AREA: Alzheimer's DiseaseREFERENCES: Buee, L. et al. Brain Res Rev 33 95 (2000); Trinczek, B. et al. Mol Biol Cell 6, 1887 (1995).
Catalog No. 50-211-0549