Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tau protein (592-597), human TFA | 2703746-44-3 | 95.17% | 868.94 | 10 MG
SDP

Catalog No. 5000423990
Encompass_Preferred

Tau protein (592-597), human TFA is a peptide fragment of human Tau protein. The dysfunction of Tau protein is involved in neurodegeneration and dementia.

  • Peptide fragment of human Tau protein.
  • Target: Tau protein.
  • Involved in neuronal signaling pathway.
  • Appearance: White to off-white solid.
  • Purity: 95.17%.
  • Soluble in H2O: ≥ 2 mg/mL (2.30 mM).

Catalog No. 50-004-23990 Supplier Medchemexpress LLC Supplier No. HYP1707A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tau protein (592-597), human TFA is a peptide fragment of human Tau protein. The dysfunction of Tau protein is involved in neurodegeneration and dementia.

  • Peptide fragment of human Tau protein.
  • Target: Tau protein.
  • Involved in neuronal signaling pathway.
  • Appearance: White to off-white solid.
  • Purity: 95.17%.
  • Soluble in H2O: ≥ 2 mg/mL (2.30 mM).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.