Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tau protein (592-597), human TFA | 2703746-44-3 | 95.17% | 868.94 | 5 MG
SDP

Catalog No. 5000424215
Encompass_Preferred

Tau protein (592-597), human TFA is a peptide fragment of human Tau protein. The dysfunction of Tau protein is involved in neurodegeneration and dementia. It is a white to off-white solid with a molecular weight of 868.94 and a chemical formula of C36H63F3N10O11. This product is for research use only.

  • Purity of 95.17%
  • Sequence is Ac-Val-Gln-Ile-Ile-Asn-Lys-NH2 (Ac-VQIINK-NH2)
  • Solubility of ≥ 2 mg/mL (2.30 mM) in H2O
  • Recommended storage for powder at -80°C for 2 years or -20°C for 1 year
  • Recommended storage for solutions at -80°C for 6 months or -20°C for 1 month (sealed, away from moisture)
  • Associated with Neuronal Signaling, Tau Protein, neurological disease, and neurodegenerative disease

Catalog No. 50-004-24215 Supplier Medchemexpress LLC Supplier No. HYP1707A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tau protein (592-597), human TFA is a peptide fragment of human Tau protein. The dysfunction of Tau protein is involved in neurodegeneration and dementia. It is a white to off-white solid with a molecular weight of 868.94 and a chemical formula of C36H63F3N10O11. This product is for research use only.

  • Purity of 95.17%
  • Sequence is Ac-Val-Gln-Ile-Ile-Asn-Lys-NH2 (Ac-VQIINK-NH2)
  • Solubility of ≥ 2 mg/mL (2.30 mM) in H2O
  • Recommended storage for powder at -80°C for 2 years or -20°C for 1 year
  • Recommended storage for solutions at -80°C for 6 months or -20°C for 1 month (sealed, away from moisture)
  • Associated with Neuronal Signaling, Tau Protein, neurological disease, and neurodegenerative disease

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.