Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tau protein (592-597), human TFA | 2703746-44-3 | 95.2% | 868.94 | 100 MG
SDP

Catalog No. 5000423989
Encompass_Preferred

Tau protein (592-597), human TFA is a peptide fragment of human Tau protein. The dysfunction of Tau protein is involved in neurodegeneration and dementia.

  • For research use only
  • Solid appearance
  • White to off-white color
  • Sequence: Ac-Val-Gln-Ile-Ile-Asn-Lys-NH2
  • Sequence shortening: Ac-VQIINK-NH2
  • Solubility in H2O: ≥ 2 mg/mL (2.30 mM)

Catalog No. 50-004-23989 Supplier Medchemexpress LLC Supplier No. HYP1707A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tau protein (592-597), human TFA is a peptide fragment of human Tau protein. The dysfunction of Tau protein is involved in neurodegeneration and dementia.

  • For research use only
  • Solid appearance
  • White to off-white color
  • Sequence: Ac-Val-Gln-Ile-Ile-Asn-Lys-NH2
  • Sequence shortening: Ac-VQIINK-NH2
  • Solubility in H2O: ≥ 2 mg/mL (2.30 mM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.