Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tau protein (592-597), human (TFA) | 2703746-44-3 | 95.2% | 868.94 | 25 MG
SDP

Catalog No. 5000426793
Encompass_Preferred

Tau protein (592-597), human TFA, is a peptide fragment of human Tau protein. The dysfunction of Tau protein is involved in neurodegeneration and dementia, making this compound relevant for neurodegenerative disease research.

  • Purity: 95.2%
  • Molecular weight: 868.94
  • Formula: C36H63F3N10O11
  • Appearance: White to off-white solid
  • Sequence: Ac-Val-Gln-Ile-Ile-Asn-Lys-NH2 (Ac-VQIINK-NH2)
  • Solubility in H2O: ≥ 2 mg/mL (2.30 mM)
  • Recommended storage for powder: -80°C for 2 years, -20°C for 1 year
  • Recommended storage in solvent: -80°C for 6 months, -20°C for 1 month (sealed, away from moisture)
  • For research use only

Catalog No. 50-004-26793 Supplier Medchemexpress LLC Supplier No. 25MGHYP1707A
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tau protein (592-597), human TFA, is a peptide fragment of human Tau protein. The dysfunction of Tau protein is involved in neurodegeneration and dementia, making this compound relevant for neurodegenerative disease research.

  • Purity: 95.2%
  • Molecular weight: 868.94
  • Formula: C36H63F3N10O11
  • Appearance: White to off-white solid
  • Sequence: Ac-Val-Gln-Ile-Ile-Asn-Lys-NH2 (Ac-VQIINK-NH2)
  • Solubility in H2O: ≥ 2 mg/mL (2.30 mM)
  • Recommended storage for powder: -80°C for 2 years, -20°C for 1 year
  • Recommended storage in solvent: -80°C for 6 months, -20°C for 1 month (sealed, away from moisture)
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.