Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tau protein (592-597), human TFA | 2703746-44-3 | 95.2% | 868.94 | 50 MG
SDP

Catalog No. 5000424446
Encompass_Preferred

Tau protein (592-597), human TFA is a peptide fragment of human Tau protein. The dysfunction of Tau protein is involved in neurodegeneration and dementia.

  • Peptide fragment of human Tau protein.
  • Involved in neurodegeneration and dementia research.
  • Purity of 95.17%.
  • Molecular weight of 868.94.
  • White to off-white solid appearance.
  • Powder storage: 2 years at -80°C, 1 year at -20°C.
  • In solvent storage: 6 months at -80°C, 1 month at -20°C (sealed storage, away from moisture).
  • Soluble in H2O at ≥ 2 mg/mL.

Catalog No. 50-004-24446 Supplier Medchemexpress LLC Supplier No. HYP1707A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tau protein (592-597), human TFA is a peptide fragment of human Tau protein. The dysfunction of Tau protein is involved in neurodegeneration and dementia.

  • Peptide fragment of human Tau protein.
  • Involved in neurodegeneration and dementia research.
  • Purity of 95.17%.
  • Molecular weight of 868.94.
  • White to off-white solid appearance.
  • Powder storage: 2 years at -80°C, 1 year at -20°C.
  • In solvent storage: 6 months at -80°C, 1 month at -20°C (sealed storage, away from moisture).
  • Soluble in H2O at ≥ 2 mg/mL.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.