Learn More
Sigma Aldrich Fine Chemicals Biosciences Tau Protein Ladder 6 isofo
Supplier:  Sigma Aldrich Fine Chemicals Biosciences T795150UL
Encompass
Tau gene spanning 100 kb with 16 exons is mapped to human chromosome 17q21. The six alternative splice variants of protein ranging in size from 352-441 amino acids have been identified in human adult brain. Tau is a member of the microtubule-associated protein (MAP) family. It is predominantly expressed in neurons.
Catalog No. 50-290-0055