Learn More
Quanterix Inc TAU SIMOA 2.0 ASSAY KIT
SDP
Catalog No. NC1060889
Encompass
    \n
  • Ultrasensitive detection for total tau measurement.
  • \n
  • Compatible with EDTA plasma, serum, and cerebrospinal fluid (CSF).
  • \n
  • Works with Quanterix Simoa SR-X, HD-1, and HD-X platforms.
  • \n
  • Detects all six human tau isoforms.
  • \n
  • Epitope located in midregion of molecule.
  • \n
  • Tests available per kit: 96.
  • \n
  • Lower limit of quantification (LLOQ) for SR-X: 0.0491 pg/mL.
  • \n
  • Lower limit of quantification (LLOQ) for HD-1/HD-X: 0.062 pg/mL.
  • \n
  • Limit of detection (LOD) for SR-X: 0.0146 pg/mL.
  • \n
  • Dynamic range for serum and plasma (SR-X): 0 - 400 pg/mL.
  • \n
  • Dynamic range for serum and plasma (HD-1/HD-X): 0 - 360 pg/mL.
  • \n
  • Dynamic range for CSF: 0 - 1000 pg/mL.
  • \n
  • Minimum required dilution: 10x to 640x.
  • \n
  • Research use only: true.
  • \n
Catalog No. NC1060889 Supplier Quanterix Inc Supplier No. 101552
    \n
  • Ultrasensitive detection for total tau measurement.
  • \n
  • Compatible with EDTA plasma, serum, and cerebrospinal fluid (CSF).
  • \n
  • Works with Quanterix Simoa SR-X, HD-1, and HD-X platforms.
  • \n
  • Detects all six human tau isoforms.
  • \n
  • Epitope located in midregion of molecule.
  • \n
  • Tests available per kit: 96.
  • \n
  • Lower limit of quantification (LLOQ) for SR-X: 0.0491 pg/mL.
  • \n
  • Lower limit of quantification (LLOQ) for HD-1/HD-X: 0.062 pg/mL.
  • \n
  • Limit of detection (LOD) for SR-X: 0.0146 pg/mL.
  • \n
  • Dynamic range for serum and plasma (SR-X): 0 - 400 pg/mL.
  • \n
  • Dynamic range for serum and plasma (HD-1/HD-X): 0 - 360 pg/mL.
  • \n
  • Dynamic range for CSF: 0 - 1000 pg/mL.
  • \n
  • Minimum required dilution: 10x to 640x.
  • \n
  • Research use only: true.
  • \n