Learn More
AnaSpec Tau (Tau-441) Protein, Fluorescently Labeled, Human Source, 50 µg
Supplier:  AnaSpec AS56085
Encompass
Introducing Tau (Tau-441), a recombinant human protein essential for neuroscience research, particularly in studies related to Alzheimer's Disease. This highly purified protein promotes neuronal microtubule assembly and stabilization, serving as a robust tool for protein aggregation and in vitro imaging applications. Labeled with HiLyte™ Fluor 488, it provides fluorescence for diverse research uses. Suitable for a wide range of experimental setups, it ensures consistency and reliability in research outcomes.
Catalog No. NC2604377