Learn More
Medchemexpress LLC Tau tracer 2 (Pl-2620) | 2173361-80-1 | 99.9% | 264.26 | C15H9FN4 | 5 MG
SDP
Catalog No. 5000222663
Encompass_Preferred
Tau tracer 2 (Pl-2620) is a small-molecule PET imaging tracer that binds aggregated Tau protein for preclinical and diagnostic research into tauopathies such as Alzheimer's disease. Supplied as an off-white to light-yellow solid, it offers high purity and established solubility and storage profiles for reliable experimental use.
  • High purity (99.92%).
  • Suitable for PET imaging of tau aggregates in preclinical studies.
  • Off-white to light-yellow solid, easy handling.
  • Documented solubility in DMSO and recommended in vivo formulations.
  • Long-term storage stability as powder at -20°C and 4°C.
  • Available in small laboratory pack sizes for research use.
Catalog No. 50-002-22663 Supplier Medchemexpress LLC Supplier No. HY1348805MG
Tau tracer 2 (Pl-2620) is a small-molecule PET imaging tracer that binds aggregated Tau protein for preclinical and diagnostic research into tauopathies such as Alzheimer's disease. Supplied as an off-white to light-yellow solid, it offers high purity and established solubility and storage profiles for reliable experimental use.
  • High purity (99.92%).
  • Suitable for PET imaging of tau aggregates in preclinical studies.
  • Off-white to light-yellow solid, easy handling.
  • Documented solubility in DMSO and recommended in vivo formulations.
  • Long-term storage stability as powder at -20°C and 4°C.
  • Available in small laboratory pack sizes for research use.