Learn More
Apexbio Technology LLC Taurine 107-35-7 1g
SDP
Catalog No. 5000088560
Encompass_Preferred
Taurine (CAS 107-35-7) is a sulfur-containing -amino acid abundantly present in animal tissues It modulates a range of physiological processes including osmoregulation calcium signaling and membrane stabilization Taurine acts as a neuromodulator in the central nervous system and influences neurotransmitter regulation ion channel function and cellular antioxidant defense mechanisms It is frequently utilized in biomedical research to investigate its roles in cardiovascular function neuroprotection and metabolic regulation as well as to explore its impact on cellular homeostasis under physiological and pathological conditions
Catalog No. 50-000-88560 Supplier Apexbio Technology LLC Supplier No. B18461000
Taurine (CAS 107-35-7) is a sulfur-containing -amino acid abundantly present in animal tissues It modulates a range of physiological processes including osmoregulation calcium signaling and membrane stabilization Taurine acts as a neuromodulator in the central nervous system and influences neurotransmitter regulation ion channel function and cellular antioxidant defense mechanisms It is frequently utilized in biomedical research to investigate its roles in cardiovascular function neuroprotection and metabolic regulation as well as to explore its impact on cellular homeostasis under physiological and pathological conditions