Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Taurine-13C2 | 70155-54-3 | MFCD01075566 | 99.9% | 5 MG
SDP

Catalog No. 5000407260
Encompass_Preferred

Taurine-13C2 is a 13C-labeled form of Taurine, a sulfur-containing amino acid and organic osmolyte. It plays a role in cell volume regulation, serves as a substrate for bile salt formation, and modulates intracellular free calcium concentration. Taurine has also been shown to activate autophagy in adipocytes.

  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Molecular weight: 127.13
  • Isotopic purity: ≥99 atom % 13C
  • Assay: ≥98% (CP)
  • Mass shift: M+2
  • Physical form: solid
  • Suitable for bio NMR and mass spectrometry (MS) techniques
  • Melting point: >300 °C

Catalog No. 50-004-07260 Supplier Medchemexpress LLC Supplier No. HYB0351S15MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Taurine-13C2 is a 13C-labeled form of Taurine, a sulfur-containing amino acid and organic osmolyte. It plays a role in cell volume regulation, serves as a substrate for bile salt formation, and modulates intracellular free calcium concentration. Taurine has also been shown to activate autophagy in adipocytes.

  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Molecular weight: 127.13
  • Isotopic purity: ≥99 atom % 13C
  • Assay: ≥98% (CP)
  • Mass shift: M+2
  • Physical form: solid
  • Suitable for bio NMR and mass spectrometry (MS) techniques
  • Melting point: >300 °C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.