Learn More
AdipoGen Taurine
SDP
Catalog No. 502559966
Encompass_Preferred
Chemical. CAS 107-35-7. Formula C2H7NO3S. MW 125.14. Synthetic. Organic acid found in animal tissues. Major constituent of bile. Non-selective endogenous agonist at glycine receptors. Has many biological roles such as conjugation of bile acids, antioxidation, osmoregulation, membrane stabilization and modulation of calcium signaling, brain development and in the enhancement of bile flow and cholesterol clearance by the liver. It is essential for cardiovascular function, and development and function of skeletal muscle, the retina and the central nervous system. Antioxidant. Inhibitor of lipid peroxidation. Shows anti-inflammatory and anti-obesity activities. Used in cosmetics due to its antifibrotic properties and maintaining skin hydration.
Catalog No. 50-255-9966 Supplier AdipoGen Supplier No. CDXT0187G500
Chemical. CAS 107-35-7. Formula C2H7NO3S. MW 125.14. Synthetic. Organic acid found in animal tissues. Major constituent of bile. Non-selective endogenous agonist at glycine receptors. Has many biological roles such as conjugation of bile acids, antioxidation, osmoregulation, membrane stabilization and modulation of calcium signaling, brain development and in the enhancement of bile flow and cholesterol clearance by the liver. It is essential for cardiovascular function, and development and function of skeletal muscle, the retina and the central nervous system. Antioxidant. Inhibitor of lipid peroxidation. Shows anti-inflammatory and anti-obesity activities. Used in cosmetics due to its antifibrotic properties and maintaining skin hydration.