Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Taurine, 99.6%, MP Biomedicals™

Catalog No. ICN10300390 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10300390 500 g
ICN10300310 10 g
ICN10300380 100 g
ICN10300391 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN10300390 Supplier MP Biomedicals, Inc Supplier No. 0210300390
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 107-35-7
Molecular Formula C2H7NO3S
Molecular Weight (g/mol) 125.14
MDL Number MFCD00008197
InChI Key XOAAWQZATWQOTB-UHFFFAOYSA-N
Synonym tetrabutylammonium chloride, tetrabutyl ammonium chloride, tetra-n-butylammonium chloride, tbac, tetrabutylazanium chloride, 1-butanaminium, n,n,n-tributyl-, chloride, tetrabutylammoniumchloride, 1-butanaminium, n,n,n-tributyl-, chloride 1:1, n,n,n-tributyl-1-butanaminium chloride, n,n,n-tributylbutan-1-aminium chloride
PubChem CID 70681
ChEBI CHEBI:51988
IUPAC Name 2-aminoethane-1-sulfonic acid
SMILES NCCS(O)(=O)=O

Specifications

Color White
Assay Percent Range ≥99%
Quantity 500 g
Formula Weight 125.15
Percent Purity 0.996
Physical Form Crystalline Powder
Chemical Name or Material Taurine
EINECSNumber 203-483-8
Recommended Storage Store at Room Temperature (15° to 30°C)
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.