Learn More
ATHENA ES Taurine, >99% pure, fermentation produced
Supplier:  ATHENA ES 11011G
Encompass_Preferred
Bio-Taurine is fermentation-produced taurine with >99% purity. It is not derived from petrochemicals, is manufactured without antibiotics, contains no ethylene oxide, and is made without animal-derived parts. Manufactured by Natáur Inc. For research use only.
Catalog No. NC3973104