Learn More
Medchemexpress LLC Taurine-d4 | 342611-14-7 | 99.96% | 1 MG
SDP
Catalog No. 5000405273
Encompass_Preferred
Taurine-d4 is the deuterium labeled Taurine. Taurine, a sulfur-containing amino acid and an organic osmolyte, is involved in cell volume regulation, provides a substrate for the formation of bile salts, and plays a role in modulating intracellular free calcium concentration. It can activate autophagy in adipocytes.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-05273 Supplier Medchemexpress LLC Supplier No. HYB0351S1MG
Taurine-d4 is the deuterium labeled Taurine. Taurine, a sulfur-containing amino acid and an organic osmolyte, is involved in cell volume regulation, provides a substrate for the formation of bile salts, and plays a role in modulating intracellular free calcium concentration. It can activate autophagy in adipocytes.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.