Learn More
Abcam Taurine, Partial glycine receptor agonist, 1G
Catalog No. 502706940
Encompass_Preferred
MW 125.15 Da, Purity >98%. Partial glycine receptor agonist. Abundant free amino acid in the retina and central brain and elicits a variety of biological effects. Able to cross the blood-brain barrier.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6940 Supplier Abcam Supplier No. AB1410631G
MW 125.15 Da, Purity >98%. Partial glycine receptor agonist. Abundant free amino acid in the retina and central brain and elicits a variety of biological effects. Able to cross the blood-brain barrier.
The product is subject to the following: Abcam Restricted Use Statement