Learn More
Cayman Chemical Tauro-α-muricholic Acid (sodium salt), Purity: ≥95%, CAS: 2260905-08-4, 1 mg
SDP
Catalog No. 502985571
Encompass_Preferred
An FXR receptor antagonist (IC50 = 28 µM); a taurine-conjugated form of the murine-specific primary bile acid α-muricholic acid
Catalog No. 50-298-5571 Supplier Cayman Chemical Supplier No. 202881 MG
An FXR receptor antagonist (IC50 = 28 µM); a taurine-conjugated form of the murine-specific primary bile acid α-muricholic acid