Learn More
Medchemexpress LLC HY-135399 1mg Medchemexpress, Tauro-Obeticholic acid CAS:863239-61-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1353991MG
Encompass_Preferred
Medchemexpress, HY-135399 1mg Tauro-Obeticholic acid CAS:863239-61-6 Tauro-Obeticholic acid is an active metabolite of Obeticholic acid. Obeticholic acid is an orally bioavailable farnesoid-X receptor (FXR) agonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1387