Learn More
Cayman Chemical Tauro-Obeticholic Acid, Purity: ≥95%, CAS: 863239-61-6, 5 mg
SDP
Catalog No. 502938555
Encompass_Preferred
An active metabolite of obeticholic acid; can be reconverted back to obeticholic acid by intestinal microflora
Catalog No. 50-293-8555 Supplier Cayman Chemical Supplier No. 282435 MG
An active metabolite of obeticholic acid; can be reconverted back to obeticholic acid by intestinal microflora