Learn More
Cayman Chemical Taurochenodeoxycholic Acid (sodium salt), Purity: ≥95%, CAS: 6009-98-9, 500 mg
SDP
Catalog No. 502984358
Encompass_Preferred
A taurine-conjugated form of CDCA; serum levels increase approximately 5-fold in within two hours and begin to decrease within four hours during an oral lipid tolerance test in humans; serum levels are increased in patients with liver cirrhosis and may serve as a marker of disease progression
Catalog No. 50-298-4358 Supplier Cayman Chemical Supplier No. 20275500 MG
A taurine-conjugated form of CDCA; serum levels increase approximately 5-fold in within two hours and begin to decrease within four hours during an oral lipid tolerance test in humans; serum levels are increased in patients with liver cirrhosis and may serve as a marker of disease progression