Learn More
Cayman Chemical TaurochenodeoxycholIc ACD 50mg
SDP
Catalog No. 502984456
Encompass_Preferred
A taurine-conjugated form of CDCA; serum levels increase approximately 5-fold in within two hours and begin to decrease within four hours during an oral lipid tolerance test in humans; serum levels are increased in patients with liver cirrhosis and may serve as a marker of disease progression
Catalog No. 50-298-4456 Supplier Cayman Chemical Supplier No. 2027550 MG
A taurine-conjugated form of CDCA; serum levels increase approximately 5-fold in within two hours and begin to decrease within four hours during an oral lipid tolerance test in humans; serum levels are increased in patients with liver cirrhosis and may serve as a marker of disease progression