Learn More
Medchemexpress LLC Taurochenodeoxycholic acid | 516-35-8 | 499.70 | 250 MG
SDP
Catalog No. 5000389460
Encompass_Preferred
Taurochenodeoxycholic acid (12-Deoxycholyltaurine) is a primary bioactive substance found in animal bile acid. It is known to induce apoptosis and exhibits significant anti-inflammatory and immune regulation properties.
  • Induces apoptosis in NR8383 cells in a concentration-dependent manner.
  • Augments PKC mRNA levels and activities.
  • Increases JNK, caspase-3, and caspase-8 mRNA expression levels and activities.
  • Decreases pulmonary coefficient and pathological damage in the lungs of model mice.
  • Reduces TNF-α and TIMP-2 expression in pulmonary tissues in pulmonary fibrosis mice.
  • Normalizes clinical inflammatory parameters and attenuates intestinal inflammation.
  • Suppresses paw swelling and polyarthritis index in AA rats.
  • Amends radiologic changes in AA rats.
  • Suppresses overproduction and mRNA expression of TNF-α, IL-1β, and IL-6 in serum and synovium tissue.
Catalog No. 50-003-89460 Supplier Medchemexpress LLC Supplier No. 250MGHYN1429
Taurochenodeoxycholic acid (12-Deoxycholyltaurine) is a primary bioactive substance found in animal bile acid. It is known to induce apoptosis and exhibits significant anti-inflammatory and immune regulation properties.
  • Induces apoptosis in NR8383 cells in a concentration-dependent manner.
  • Augments PKC mRNA levels and activities.
  • Increases JNK, caspase-3, and caspase-8 mRNA expression levels and activities.
  • Decreases pulmonary coefficient and pathological damage in the lungs of model mice.
  • Reduces TNF-α and TIMP-2 expression in pulmonary tissues in pulmonary fibrosis mice.
  • Normalizes clinical inflammatory parameters and attenuates intestinal inflammation.
  • Suppresses paw swelling and polyarthritis index in AA rats.
  • Amends radiologic changes in AA rats.
  • Suppresses overproduction and mRNA expression of TNF-α, IL-1β, and IL-6 in serum and synovium tissue.