Learn More
American Radiolabeled Chemicals Inc TAUROCHOLIC AC 14C 50UC
SDP
Supplier:  American Radiolabeled Chemicals Inc ARC 348950UC
Encompass_Preferred
Taurocholic acid-taurine-1,2-14C 50uCi in ethanol, 0.1 mCi/ml, S.A. 50-60 mCi/mmol, M.W. 515.7, Shipped in Dry Ice, Exclusive
Catalog No. 50-901-288