Learn More
Cayman Chemical TaurocholIc ACD MaxSpec Sta
SDP
Catalog No. 502990938
Encompass_Preferred
A quantitative analytical standard guaranteed to meet MaxSpec identity, purity, stability, and concentration specifications
Catalog No. 50-299-0938 Supplier Cayman Chemical Supplier No. 31374100 UG
Add to Cart
Add to Cart
A quantitative analytical standard guaranteed to meet MaxSpec identity, purity, stability, and concentration specifications