Learn More
Cayman Chemical Taurocholic Acid 3-sulfate (sodium salt), Purity: ≥95%, CAS: 71781-33-4, 10 mg
SDP
Catalog No. 502895122
Encompass_Preferred
A metabolite of taurocholic acid; plasma levels are elevated in wild-type and Sort1 knockout mice at 6 hours post-BDL and are further elevated in Sort1 knockout mice at 24 hours post-BDL
Catalog No. 50-289-5122 Supplier Cayman Chemical Supplier No. 900253810 MG
A metabolite of taurocholic acid; plasma levels are elevated in wild-type and Sort1 knockout mice at 6 hours post-BDL and are further elevated in Sort1 knockout mice at 24 hours post-BDL