Learn More
Medchemexpress LLC Taurocholic acid-d4 | 252030-90-3 | 99.5% | 100 UG
SDP
Catalog No. 5000412865
Encompass_Preferred
Taurocholic acid-d4 is deuterium labeled Taurocholic acid, a bile acid involved in the emulsification of fats. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium labeled for use as a tracer
  • Involved in fat emulsification
  • Suitable for quantitative analysis
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-12865 Supplier Medchemexpress LLC Supplier No. HYB1788S1100UG
Taurocholic acid-d4 is deuterium labeled Taurocholic acid, a bile acid involved in the emulsification of fats. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium labeled for use as a tracer
  • Involved in fat emulsification
  • Suitable for quantitative analysis
  • Potential to affect pharmacokinetic and metabolic profiles of drugs