Learn More
Medchemexpress LLC Taurocholic acid-d4 (N-Choloyltaurine-d4) | 252030-90-3 | 99.5% | 500 UG
SDP
Catalog No. 5000411720
Encompass_Preferred
Taurocholic acid-d4 is a deuterium-labeled form of Taurocholic acid, a bile acid involved in the emulsification of fats. This compound is used as a tracer and as an internal standard for quantitative analysis in drug development processes where stable heavy isotopes are incorporated into drug molecules. Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer.
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes into drug molecules for quantitation.
  • Potential to affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-11720 Supplier Medchemexpress LLC Supplier No. HYB1788S500UG
Taurocholic acid-d4 is a deuterium-labeled form of Taurocholic acid, a bile acid involved in the emulsification of fats. This compound is used as a tracer and as an internal standard for quantitative analysis in drug development processes where stable heavy isotopes are incorporated into drug molecules. Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer.
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes into drug molecules for quantitation.
  • Potential to affect pharmacokinetic and metabolic profiles of drugs.