Learn More
Medchemexpress LLC Taurocholic acid-d4 sodium | 2410279-93-3 | 98.2% | 1 MG
SDP
Catalog No. 5000412826
Encompass_Preferred
Taurocholic acid-d4 (sodium) is the deuterium-labeled version of Taurocholic acid, which is a bile acid involved in the emulsification of fats. It is intended for research use only and is not sold to patients. This compound can be utilized as a tracer and as an internal standard for quantitative analysis through methods like NMR, GC-MS, or LC-MS. Deuteration, the incorporation of stable heavy isotopes like hydrogen and carbon into drug molecules, is used as a tracer for quantitation during drug development and has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled Taurocholic acid.
  • Involved in the emulsification of fats.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-12826 Supplier Medchemexpress LLC Supplier No. HYB1788S1MG
Taurocholic acid-d4 (sodium) is the deuterium-labeled version of Taurocholic acid, which is a bile acid involved in the emulsification of fats. It is intended for research use only and is not sold to patients. This compound can be utilized as a tracer and as an internal standard for quantitative analysis through methods like NMR, GC-MS, or LC-MS. Deuteration, the incorporation of stable heavy isotopes like hydrogen and carbon into drug molecules, is used as a tracer for quantitation during drug development and has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled Taurocholic acid.
  • Involved in the emulsification of fats.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.