Learn More
Medchemexpress LLC Taurocholic acid-d4 sodium (N-Choloyltaurine-d4 sodium) | 2410279-93-3 | 98.2% | 50 MG
SDP
Catalog No. 5000411525
Encompass_Preferred
Taurocholic acid-d4 (sodium) is a deuterium-labeled version of Taurocholic acid. Taurocholic acid (N-Choloyltaurine) is a bile acid that helps in the emulsification of fats.
  • Deuterium labeled
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-11525 Supplier Medchemexpress LLC Supplier No. HYB1788S50MG
Taurocholic acid-d4 (sodium) is a deuterium-labeled version of Taurocholic acid. Taurocholic acid (N-Choloyltaurine) is a bile acid that helps in the emulsification of fats.
  • Deuterium labeled
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS