Learn More
Cayman Chemical TaurocholIc AcId-d4sodIM500ug
SDP
Catalog No. 502987164
Encompass_Preferred
An internal standard for the quantification of taurocholic acid by GC- or LC-MS
Catalog No. 50-298-7164 Supplier Cayman Chemical Supplier No. 21891500 UG
An internal standard for the quantification of taurocholic acid by GC- or LC-MS