Learn More
Medchemexpress LLC Taurocholic acid (N-Choloyltaurine) | 81-24-3 | 99.8% | 50 MG
SDP
Catalog No. 5000412661
Encompass_Preferred
Taurocholic acid (N-Choloyltaurine) exhibits notable bioactive effects, including an inhibitory potential against hepatic artery ligation-induced biliary damage through the upregulation of VEGF-A expression. It also possesses an immunoregulation effect.
  • Inhibits hepatic artery ligation-induced biliary damage
  • Upregulates VEGF-A expression
  • Has immunoregulation effect
  • For research use only
Catalog No. 50-004-12661 Supplier Medchemexpress LLC Supplier No. HYB178850MG
Add to Cart
Add to Cart
Taurocholic acid (N-Choloyltaurine) exhibits notable bioactive effects, including an inhibitory potential against hepatic artery ligation-induced biliary damage through the upregulation of VEGF-A expression. It also possesses an immunoregulation effect.
  • Inhibits hepatic artery ligation-induced biliary damage
  • Upregulates VEGF-A expression
  • Has immunoregulation effect
  • For research use only