Learn More
Cayman Chemical Taurocholic Acid (sodium salt), 145-42-6, 5 g
SDP
Supplier:  Cayman Chemical 162155
Encompass_Preferred
A bile salt formed in the liver by conjugation of cholic acid with taurine that can be used to solubilize lipids and membrane-bound proteins
Catalog No. NC1380829