Learn More
Selleck Chemical LLC Taurocholic acid sodium salt
SDP
Supplier:  Selleck Chemical LLC E01541G
Encompass_Preferred
Taurocholic acid sodium salt (Sodium Taurocholate TANa) is a sodium salt of taurocholic acid and occurs in the bile of mammals Taurocholic acid is used as a cholagogue and choleretic
Catalog No. 50-313-6880