Learn More
Medchemexpress LLC Taurocholic acid sodium salt hydrate | 345909-26-4 | 98.5% | 500 MG
SDP
Catalog No. 5000413495
Encompass_Preferred
Taurocholic acid sodium salt hydrate is a chemical compound with a molecular formula of C26H46NNaO8S and a molecular weight of 555.70. It is typically supplied as a white to yellow solid.
  • Consistent with structure based on 1H NMR spectrum
  • High purity of 98.51% confirmed by HPLC
  • Complies with given specifications
Catalog No. 50-004-13495 Supplier Medchemexpress LLC Supplier No. HYB1131500MG
Taurocholic acid sodium salt hydrate is a chemical compound with a molecular formula of C26H46NNaO8S and a molecular weight of 555.70. It is typically supplied as a white to yellow solid.
  • Consistent with structure based on 1H NMR spectrum
  • High purity of 98.51% confirmed by HPLC
  • Complies with given specifications