Learn More
Cayman Chemical Taurocholic Acid (sodium salt), Purity: ≥95%, CAS: 145-42-6, 25 g
SDP
Catalog No. 502971304
Encompass_Preferred
A taurine-conjugated form of cholic acid; serum levels of TCA are decreased in patients with Crohn’s disease and those with ulcerative colitis with no extraintestinal manifestations, but are increased in patients with ulcerative colitis accompanied by hepatobiliary disease
Catalog No. 50-297-1304 Supplier Cayman Chemical Supplier No. 1621525 G
A taurine-conjugated form of cholic acid; serum levels of TCA are decreased in patients with Crohn’s disease and those with ulcerative colitis with no extraintestinal manifestations, but are increased in patients with ulcerative colitis accompanied by hepatobiliary disease