Learn More
Medchemexpress LLC HY-B1899A 10mg Medchemexpress, Taurodeoxychloic Acid (sodium hydrate) CAS:110026-03-4 Purity:>98%
SDP
Catalog No. 501933032
Encompass_Preferred
Medchemexpress, HY-B1899A 10mg Taurodeoxychloic Acid (sodium hydrate) CAS:110026-03-4 Taurodeoxychloic Acid (sodium hydrate) prevents apoptosis by blocking a calcium-mediated apoptotic pathway as well as caspase-12 activation. Taurodeoxychloic Acid (sodium hydrate) is investigated for use in several conditions such as Primary Biliary Cirrhosis (PBC), insulin resistance, amyloidosis, Cystic Fibrosis, Cholestasis, and Amyotrophic Lateral Sclerosis [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3032 Supplier Medchemexpress LLC Supplier No. HYB1899A10MG
Medchemexpress, HY-B1899A 10mg Taurodeoxychloic Acid (sodium hydrate) CAS:110026-03-4 Taurodeoxychloic Acid (sodium hydrate) prevents apoptosis by blocking a calcium-mediated apoptotic pathway as well as caspase-12 activation. Taurodeoxychloic Acid (sodium hydrate) is investigated for use in several conditions such as Primary Biliary Cirrhosis (PBC), insulin resistance, amyloidosis, Cystic Fibrosis, Cholestasis, and Amyotrophic Lateral Sclerosis [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.