Learn More
Medchemexpress LLC HY-128853 500mg Medchemexpress, Taurodeoxycholate (sodium salt) CAS:1180-95-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY128853500MG
Encompass_Preferred
Medchemexpress, HY-128853 500mg Taurodeoxycholate (sodium salt) CAS:1180-95-6 Taurodeoxycholate sodium salt is a bile salt-related anionic detergent used for isolation of membrane proteins including inner mitochondrial membrane proteins. Taurodeoxycholate (TDCA) inhibits various inflammatory responses [2][3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0137