Learn More
Medchemexpress LLC Taurodeoxycholic acid | 516-50-7 | 98.0% | 50 MG
SDP
Catalog No. 5000409917
Encompass_Preferred
Taurodeoxycholate sodium salt is a bile salt-related anionic detergent. Taurodeoxycholic acid is formed in the liver by the conjugation of deoxycholate with Taurine. It is used for isolating membrane proteins, including inner mitochondrial membrane proteins. Taurodeoxycholic acid (TDCA) has anti-inflammatory and neuroprotective effects.
  • Exhibits anti-inflammatory effects
  • Exhibits neuroprotective effects
  • Ameliorates dextran sodium sulfate (DSS)-induced colitis in mice
  • Prevents neuropathology and associated behavioral deficits in the 3-nitropropionic acid rat model of Huntington's disease (HD)
  • Leads to a significant reduction in striatal neuropathology of the R6/2 transgenic HD mouse
  • Confers protection to C57BL/6N mice with sepsis
Catalog No. 50-004-09917 Supplier Medchemexpress LLC Supplier No. HYB189950MG
Add to Cart
Add to Cart
Taurodeoxycholate sodium salt is a bile salt-related anionic detergent. Taurodeoxycholic acid is formed in the liver by the conjugation of deoxycholate with Taurine. It is used for isolating membrane proteins, including inner mitochondrial membrane proteins. Taurodeoxycholic acid (TDCA) has anti-inflammatory and neuroprotective effects.
  • Exhibits anti-inflammatory effects
  • Exhibits neuroprotective effects
  • Ameliorates dextran sodium sulfate (DSS)-induced colitis in mice
  • Prevents neuropathology and associated behavioral deficits in the 3-nitropropionic acid rat model of Huntington's disease (HD)
  • Leads to a significant reduction in striatal neuropathology of the R6/2 transgenic HD mouse
  • Confers protection to C57BL/6N mice with sepsis