Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Taurohyocholic acid | 32747-07-2 | 99.7% | 5 MG
SDP

Catalog No. 5000230457
Encompass_Preferred

Taurohyocholic acid is a taurine-conjugated hyocholic bile acid provided as a high-purity research reagent (CAS 32747-07-2). It is intended for in vitro biochemical and analytical studies of bile acid metabolism and signaling.

  • High purity: 99.69% (reported)
  • Chemical formula C26H45NO7S
  • Molecular weight 515.70 g/mol
  • Available in small-mass quantities suitable for analytical work
  • Soluble in DMSO for preparation of stock solutions

Catalog No. 50-002-30457 Supplier Medchemexpress LLC Supplier No. HY1217995MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Taurohyocholic acid is a taurine-conjugated hyocholic bile acid provided as a high-purity research reagent (CAS 32747-07-2). It is intended for in vitro biochemical and analytical studies of bile acid metabolism and signaling.

  • High purity: 99.69% (reported)
  • Chemical formula C26H45NO7S
  • Molecular weight 515.70 g/mol
  • Available in small-mass quantities suitable for analytical work
  • Soluble in DMSO for preparation of stock solutions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.