Learn More
Cayman Chemical Taurohyocholic Acid (sodium salt), Purity: ≥95%, CAS: 117997-17-8, 25 mg
SDP
Catalog No. 502994869
Encompass_Preferred
A taurine-conjugated form of hyocholic acid; inhibits precipitation of cholesterol crystals by stabilizing cholesterol in the liquid-crystalline phase; prevents cholestasis and cellular necrosis induced by taurolithocholic acid in isolated rat livers; levels are increased in the urine of patients with hepatitis B-induced cirrhosis
Catalog No. 50-299-4869 Supplier Cayman Chemical Supplier No. 2266925 MG
A taurine-conjugated form of hyocholic acid; inhibits precipitation of cholesterol crystals by stabilizing cholesterol in the liquid-crystalline phase; prevents cholestasis and cellular necrosis induced by taurolithocholic acid in isolated rat livers; levels are increased in the urine of patients with hepatitis B-induced cirrhosis