Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Taurohyodeoxycholic 10mg | 386523 | 10 MG
SDP

Catalog No. 5000228102
Encompass_Preferred

Taurohyodeoxycholic acid is a taurine-conjugated 6α-hydroxylated bile acid used in biochemical and preclinical research to investigate liver and intestinal inflammation. It modulates inflammatory markers such as MPO, TNF-α, IL-6, and COX-2, and is applied in models of nonalcoholic fatty liver disease, colitis, and biliary disorders.

  • Purity: 99.7%
  • Molecular weight: 499.7 g·mol⁻¹
  • Chemical formula: C26H45NO6S
  • Available as small solid aliquots (for example 10 MG) and as DMSO solution (10 mM x 1 mL)
  • Suitable for in vitro assays and in vivo disease models
  • Datasheet and SDS available for handling and safety information

Catalog No. 50-002-28102 Supplier Medchemexpress LLC Supplier No. HY11436010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Taurohyodeoxycholic acid is a taurine-conjugated 6α-hydroxylated bile acid used in biochemical and preclinical research to investigate liver and intestinal inflammation. It modulates inflammatory markers such as MPO, TNF-α, IL-6, and COX-2, and is applied in models of nonalcoholic fatty liver disease, colitis, and biliary disorders.

  • Purity: 99.7%
  • Molecular weight: 499.7 g·mol⁻¹
  • Chemical formula: C26H45NO6S
  • Available as small solid aliquots (for example 10 MG) and as DMSO solution (10 mM x 1 mL)
  • Suitable for in vitro assays and in vivo disease models
  • Datasheet and SDS available for handling and safety information

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.