Learn More
Medchemexpress LLC Taurohyodeoxycholic 50mg | 38411-85-7 | 50 MG
SDP
Catalog No. 5000229652
Encompass_Preferred
Taurohyodeoxycholic acid is an orally active 6α-hydroxylated bile acid used in preclinical research to modulate inflammatory responses and to study liver and intestinal disease models.
  • Orally active 6α-hydroxylated bile acid used in biological studies.
  • Modulates inflammatory markers such as MPO, TNF-α, and IL-6.
  • High purity (99.7%) suitable for in vitro and in vivo assays.
  • Soluble in DMSO at 100 mg/mL; multiple in vivo formulation protocols available.
  • Stable when stored as powder at -20°C for extended periods.
Catalog No. 50-002-29652 Supplier Medchemexpress LLC Supplier No. HY114360A50MG
Taurohyodeoxycholic acid is an orally active 6α-hydroxylated bile acid used in preclinical research to modulate inflammatory responses and to study liver and intestinal disease models.
  • Orally active 6α-hydroxylated bile acid used in biological studies.
  • Modulates inflammatory markers such as MPO, TNF-α, and IL-6.
  • High purity (99.7%) suitable for in vitro and in vivo assays.
  • Soluble in DMSO at 100 mg/mL; multiple in vivo formulation protocols available.
  • Stable when stored as powder at -20°C for extended periods.