Learn More
Medchemexpress LLC Taurohyodeoxycholic 50mg | 386523 | 50 MG
SDP
Catalog No. 5000229836
Encompass_Preferred
Taurohyodeoxycholic acid is a taurine-conjugated 6α-hydroxylated bile acid used in preclinical research to study liver disease, colitis, and biliary disorders. It exhibits anti-inflammatory effects, including reductions in MPO activity, TNF-α, and IL-6.
  • Used to study nonalcoholic fatty liver disease and colitis.
  • Shows anti-inflammatory activity in animal models.
  • Available in multiple solid and solution pack sizes.
  • Chemical formula C26H45NO6S; molecular weight 499.70 g/mol.
Catalog No. 50-002-29836 Supplier Medchemexpress LLC Supplier No. HY11436050MG
Taurohyodeoxycholic acid is a taurine-conjugated 6α-hydroxylated bile acid used in preclinical research to study liver disease, colitis, and biliary disorders. It exhibits anti-inflammatory effects, including reductions in MPO activity, TNF-α, and IL-6.
  • Used to study nonalcoholic fatty liver disease and colitis.
  • Shows anti-inflammatory activity in animal models.
  • Available in multiple solid and solution pack sizes.
  • Chemical formula C26H45NO6S; molecular weight 499.70 g/mol.