Learn More
Cayman Chemical Taurohyodeoxycholic Acid (sodium salt), Purity: ≥95%, CAS: 38411-85-7, 10 mg
SDP
Catalog No. 502985728
Encompass_Preferred
A taurine-conjugated form of hyodeoxycholic acid; decreases the size and weight of human gallstones in vitro; prevents hepatotoxicity induced by TCDCA, increasing bile flow as well as biliary acid and phospholipid secretion in rats; reduces myeloperoxidase activity, expression of TNF-α and IL-6, and colonic damage in a mouse model of TNBS-induced ulcerative colitis
Catalog No. 50-298-5728 Supplier Cayman Chemical Supplier No. 2195610 MG
A taurine-conjugated form of hyodeoxycholic acid; decreases the size and weight of human gallstones in vitro; prevents hepatotoxicity induced by TCDCA, increasing bile flow as well as biliary acid and phospholipid secretion in rats; reduces myeloperoxidase activity, expression of TNF-α and IL-6, and colonic damage in a mouse model of TNBS-induced ulcerative colitis