Learn More
Cayman Chemical Tauroldn 10mg
SDP
Catalog No. 502970737
Encompass_Preferred
A synthetic taurine analog with broad bactericidal and fungicidal activity; binds to the extracellular wall of bacteria, blocking adherence to epithelial and fibroblast cells; induces autophagy, apoptosis, and necrosis in human cancer cells
Catalog No. 50-297-0737 Supplier Cayman Chemical Supplier No. 1537410 MG
A synthetic taurine analog with broad bactericidal and fungicidal activity; binds to the extracellular wall of bacteria, blocking adherence to epithelial and fibroblast cells; induces autophagy, apoptosis, and necrosis in human cancer cells